ContactUsLinkComponent
Sequencing the Human Microbiome
Sequencing the Human Microbiome
# de catalogue 470126-538
Fournisseur:  Edvotek
SEQUENCING THE HUMAN MICROBIOME
Sequencing the Human Microbiome
# de catalogue 470126-538
Fournisseur:  Edvotek
Spécifications
  • Description:
    Sequencing the Human Microbiome Kit
  • No. of Lab Groups:
    10
  • Cat. no.:
    470126-538
  • Run Time:
    45 min
  • Supplier No.:
    339
Spécifications
A propos de cet article
Learn the basics of DNA sequencing and sequence homology with this lab activity.
  • No sophisticated lab equipment needed
  • Provides enough material for 10 lab groups
  • Takes 45 minutes to complete
Humans live in a delicate balance with the microorganisms that live in and on their bodies. If this balance is disrupted, harmful bacteria can multiply and cause disease. In this experiment, students will read DNA sequence obtained from automated DNA sequencing technologies. The data will be analyzed using publicly available databases to identify the bacterial species present in a patient sample. The results will be used to make a diagnosis.
This experiment contains a total of twelve sections of automated DNA sequence printouts. Students can use any DNA sequence database to perform the activities in this lab. For purposes of simplification we have chosen to illustrate the database offered by the NCBI.
Ordering information: To complete this activity students will need a computer with internet access.
Customers Who Bought This Also Bought