Semi Micro Test Tube Brush
# de catalogue 470175-996
Fournisseur: Mill-Rose
Spécifications
- Description:Semimicro Test Tube Brush
- Cat. no.:470175-996
- Supplier No.:73200B
Spécifications
A propos de cet article
Excellent for cleaning semimicro tubes and other small apparatus. Has black nylon bristles with fan style tip mounted in twisted galvanized wire. Brush area is 1.5 cm in diameter by 7 cm in length; overall length is 15 cm. Package of 12.
Customers Who Bought This Also Bought