Seines
Fournisseur: DOUGLAS NET COMPANY
Certificats
A propos de cet article
Great for Stream Sampling
- Large Area Screen for Sampling
- Collect More Fish and Specimens
Suitable for both saltwater and freshwater, this seine features 2 mm mesh and includes weights and floats. Size: 1m x 7.6m.
Customers Who Bought This Also Bought