GEO-logic Sedimentary Rocks Thin Section Topic Set
# de catalogue 470025-356
Fournisseur: Avantor
Spécifications
- Dimensions:27 x 46 mm
- No. of Slides:12
- Description:Sedimentary Rocks Thin Sections
- Cat. no.:470025-356
- Supplier No.:WARD470025-356
Spécifications
A propos de cet article
Examine the Mineral Composition of Sedimentary Rocks
- Developed jointly by experienced educators and Ward's® staff geologists
- Classroom tested for dependability
- Designed to save valuble planning and preparation time
Students can study the composition and learn the origin of sedimentary rocks when they examine the thin sections slides in the GEO-logic set. The microscope slides correspond to the rock samples in the Sedimentary Rocks Activity Set. A teacher's guide and student copymasters are included for the following activities: recognizing origin in thin sections, identifying and comparing the layering in sedimentary rock, and estimating percentages of minerals. An optical properties chart is included, and the set is housed in a hinged storage box.
Ordering information: Contents include 12 thin section glass microscope slides (27 x 46 mm), Magnifier, Teacher's Guide, and Student Copymasters.
Customers Who Bought This Also Bought