Screw Caps for Specimen Jars
Fournisseur: EMPIRE/EMCO
Certificats
A propos de cet article
These steel caps fit flint glass, screw cap wide mouth specimen jars. They are finished in black enamel and have a chemically resistant inner liner.
Customers Who Bought This Also Bought