Boreal Advanced Compound Microscope
Supplier: VWR
Certificates
About this item
Exceptional Quality at an Outstanding Value
- Rack-and-Pinion Focus Guide
- 360° Rotatable Head for Viewing from any Angle
- Long-Lasting LED Illumination
- Height: 141/2"
- Lifetime Manufacturer's Warranty
Multiple features make this a practical choice for the classroom including 4x, 10x, 40x and 100x objectives, built-in mechanical stage and corded or cordless LED illumination. Designed to withstand years of wear and tear, it features all metal construction. A reinforced joint between head and eyetube prevent stripping and recessed set screws prevent tampering. Includes Boreal Lifetime Warranty.
Two people can view a specimen simultaneously, or one viewer can add a micro-video camera with the two 10x wide-field eyepieces on this version of the high-quality scope.
Ordering information: This product includes a manufacturer's warranty.
Specifications
- Catalog No:
- 470145-732
- 470145-590
- 470177-238
- Objectives:
- 4×, 10×, 40×, 100×
- 4×, 10×, 40×, 100×
- 4×, 10×, 40×, 100×
- Stage:
- Built-In Mechanical
- Built-In Mechanical
- Built-In Mechanical