Ward’s® Shapes of Molecules Molecular Model Set, 74 Pieces
# de catalogue 470151-960
Fournisseur: VWR International
Spécifications
- Description:Shapes of Molecules Set, 74 Pieces
- Cat. no.:470151-960
- Supplier No.:SET00613
Spécifications
A propos de cet article
Create Eight VSEPR Shapes
Demonstrate VSEPR theory used to predict the shape of molecules with this complete set that contains enough parts to build eight atomic models, each with a different shape. Includes carbon, oxygen, nitrogen, sulfur, phosphorus, chlorine, boron, beryllium, chlorine, fluorine, and hydrogen elements with six lone pair orbitals and six pear-shaped orbitals. Also contains 26 pear-shaped sigma bonds and six short bonds.
Contents:
Qty Element Holes
1 Carbon 4
1 Oxygen 4
1 Nitrogen 4
1 Sulfur 6
1 Phosphorus 5
1 Chlorine 4
1 Boron 3
1 Beryllium 2
7 Chlorine 1
9 Fluorine 1
13 Hydrogen 1
6 Circular Lone Pair Orbitals
6 Pear Shaped Lone Pair Orbitals
26 Pear Shaped Sigma bonds
6 Short Bonds
Customers Who Bought This Also Bought