Ward's® Safety Signs, KELLER BROS. & MILLER, INC.
# de catalogue 470016-856
Fournisseur: KELLER BROS. & MILLER, INC.
Spécifications
- Description:Ward's Chemistry Safety Signs, Set of 9
- Cat. no.:470016-856
- Dimensions:11×6" - 20×5"
- Supplier No.:9499620
Spécifications
A propos de cet article
These signs provide helpful reminders to promote safety in the laboratory.
- Signs Label Important Safety Features in Laboratories or Classrooms
- Various Sizes
- Laminated for Durability
- Brightly Colored
These brightly colored signs communicate where safety equipment is in your laboratory or classroom.
Large, bold letters identify the location of fire blankets, eyewash stations, first aid kits, and more, so students can easily locate these apparatus in case of emergency. All signs are laminated for durability.
Ordering information: The lab safety signs include Eyewash Station, Safety Shower Station, Emergency Fire Blanket, Wear Your Safety Goggles, Fire Extinguisher, MSDS, Do Not Enter, Authorized Personnel Only, and First Aid Kit.
Customers Who Bought This Also Bought