Organic Stereo Chemistry Molecular Model Set, 365 Pieces
# de catalogue 470128-684
Fournisseur: VWR International
Spécifications
- Description:Organic Stereo Set, 364 Pieces
- Cat. no.:470128-684
- Supplier No.:SET00612
Spécifications
A propos de cet article
Designed to introduce stereochemistry in high school and introductory college chemistry courses.
- Contains 170 colored spheres representing the elements
- Contains 365 total parts
- Includes 12 orbitals
Begin the discussion of stereochemistry with this comprehensive kit that contains orbitals to allow understanding of the spatial relationship of atoms within molecules. Ideal for high school and introductory college chemistry, this set contains 170 colored spheres representing carbon, hydrogen, nitrogen, oxygen, sulfur, phosphorus, fluorine, bromine, iodine and metal elements with 12 orbitals. Diameter of spheres ranges from 15 to 22 mm.
Ordering information: Set contains the following elements (Qty, Element, Color, Holes, Dia.):
32, Carbon, Black, 3, 22 mm
12, Carbon, Black, 3, 22 mm
60, Hydrogen, White, 1, 15 mm
6, Hydrogen, White, 2, 15 mm
8, Nitrogen, Blue, 4, 22 mm
4, Nitrogen, Blue, 5, 22 mm
12, Oxygen, Red, 4, 22 mm
4, Oxygen, Red, 1, 15 mm
8, Sulfur, Yellow, 2, 22 mm
4, Sulfur, Yellow, 4, 22 mm
2, Phosphorous, Purple, 5, 22 mm
3, Phosphorous, Purple, 4, 22 mm
8, Fluorine, Pale Green, 1, 15 mm
2, Bromine, Orange, 1, 15 mm
2, Iodine, Purple, 1, 15 mm
1, Metal, Gray, 4, 22 mm
1, Metal, Gray, 5, 22 mm
1, Metal, Gray, 6, 22 mm
Set also contains 6 pink orbitals (2D), 6 gray orbitals (2D), 50 gray bonds, 12 gray long bonds, 120 white short bonds and the link remover tool.
32, Carbon, Black, 3, 22 mm
12, Carbon, Black, 3, 22 mm
60, Hydrogen, White, 1, 15 mm
6, Hydrogen, White, 2, 15 mm
8, Nitrogen, Blue, 4, 22 mm
4, Nitrogen, Blue, 5, 22 mm
12, Oxygen, Red, 4, 22 mm
4, Oxygen, Red, 1, 15 mm
8, Sulfur, Yellow, 2, 22 mm
4, Sulfur, Yellow, 4, 22 mm
2, Phosphorous, Purple, 5, 22 mm
3, Phosphorous, Purple, 4, 22 mm
8, Fluorine, Pale Green, 1, 15 mm
2, Bromine, Orange, 1, 15 mm
2, Iodine, Purple, 1, 15 mm
1, Metal, Gray, 4, 22 mm
1, Metal, Gray, 5, 22 mm
1, Metal, Gray, 6, 22 mm
Set also contains 6 pink orbitals (2D), 6 gray orbitals (2D), 50 gray bonds, 12 gray long bonds, 120 white short bonds and the link remover tool.
Customers Who Bought This Also Bought