Ward's® Laboratory Safety Poster Set
# de catalogue 470016-858
Fournisseur: KELLER BROS. & MILLER, INC.
Spécifications
- Length:30"
- Description:Ward's Chemistry Laboratory Safety Posters, Set of 7
- Height:20"
- Cat. no.:470016-858
- Supplier No.:9499625
Spécifications
A propos de cet article
Ward's Laboratory safety poster set establishes rules in the lab.
- Topics include wafting, wearing goggles, and more
- Colorful illustrations
Chart topics include wafting, combining an acid with water, tying up hair, safe placement of a Bunsen burner, safely transporting chemicals within the lab, wearing goggles, and appropriate footwear.
Each colorful poster in this series illustrates a different safety guideline for the laboratory.
Customers Who Bought This Also Bought