Chemistry Gram Stain Kit
Supplier: Ward's Science
Certificates
About this item
The Chemistry Gram Stain Kit makes gram straining easy and fast.
- Shelf Life of 36 Months
- Students Compare Microorganisms and Their Characteristics
- No Chemical Mixing
- Multiple Solutions Included
- Fast and Simple Process
It lets you compare microorganisms and their characteristics and doesn't require chemical mixing.
Certifications: Note:This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Ordering information: Kit No. 470010-662 contains
30ml crystal violet
30ml iodine potassium iodide
30ml safranin O
100ml 95% ethanol
Kit No. 470302-350 contains
500ml crystal violet
500ml iodine potassium iodide
500ml safranin O
2 X 1000ml 95% ethanol
30ml crystal violet
30ml iodine potassium iodide
30ml safranin O
100ml 95% ethanol
Kit No. 470302-350 contains
500ml crystal violet
500ml iodine potassium iodide
500ml safranin O
2 X 1000ml 95% ethanol
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.