Spécifications
- Description:Biochemistry Student Set, 135 Pieces
- Cat. no.:470149-216
- Supplier No.:SET00610
Spécifications
A propos de cet article
Adds Visual Impact to Biochemistry Demonstrations
Elements most commonly used in biochemistry are featured in this colorful, hands-on molecule building kit. Ideal for skills demonstration and practice for high school and college biochemistry students, this kit includes 79 color-coded spheres featuring carbon, oxygen, hydrogen, nitrogen, sulfur, and phosphorus elements, with holes for inserting links to represent chemical bonds. Forty-four medium bonds and twelve long bonds are included. Spheres range in diameter from 15mm to 22mm.
Contents:
Qty Element Color Holes Diameter
14 Carbon Black 4 22 mm
10 Carbon Black 3 22 mm
12 Oxygen Red 2 20 mm
2 Oxygen Red 1 15 mm
25 Hydrogen White 1 15 mm
3 Hydrogen White 2 15 mm
3 Nitrogen Blue 4 22 mm
2 Nitrogen Blue 2 20 mm
6 Nitrogen Blue 3 22 mm
1 Sulfur Yellow 2 22 mm
1 Phosphorus Purple 4 22 mm
44 Medium Bonds Grey
12 Long Bonds Grey