Ward’s® Atomic Orbital Molecular Model Set
# de catalogue 470151-958
Fournisseur: VWR International
Spécifications
- Description:Atomic Orbital Set
- Supplier No.:SET00614
Spécifications
A propos de cet article
Build 14 Atomic Orbitals
Students will discover and learn about atomic shape with this complete, colorful model set with materials to build 14 orbitals. Both P and D atomic orbitals contain pink and purple pear-shaped lobes, representing the positive and negative charges contained within each orbital. Mounted on a transparent base, this model is ideal for group demonstration and classroom display.
Customers Who Bought This Also Bought