Schistosoma japonicum, Male/Female Slide
# de catalogue 470182-034
Fournisseur: Avantor
Spécifications
- Description:Schistosoma japonicum, Male/Female Slide
- Cat. no.:470182-034
- Stain:Grenacher's borax carmine
- Supplier No.:WARD470182-034
- Orientation:Whole mount
Spécifications
A propos de cet article
Human infectious parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
Blood fluke that infects the mesenteric veins of humans. Demonstrates Male and female adults in copula.
Customers Who Bought This Also Bought