ContactUsLinkComponent
Rubber Laboratory Apron
Rubber Laboratory Apron
# de catalogue 470148-648
Fournisseur:  VWR International
APRON RUBBER ADULT 69CMX107CM
Rubber Laboratory Apron
# de catalogue 470148-648
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Garment application:
    Chemical protection
  • Gender:
    Unisex
  • Color:
    Black
  • L×W:
    42×27"
  • Fabric:
    Extra-heavy rubber and cotton tabs
  • Cat. no.:
    470148-648
  • Type:
    Rubber apron
  • Supplier No.:
    RBAP-AD
Spécifications
A propos de cet article
Durable, lightweight, rubber apron resistant to chemicals, acids, and caustics.
  • Chemical-resistant
  • Extra-heavy rubber material
  • Lightweight
  • Contains latex
Extra-heavy rubber coated sheeting plus cotton tabs on the waist and neck have made these aprons a durable lab standard for years. They resist most chemicals, acids, and caustics, yet are still lightweight.
Customers Who Bought This Also Bought