ContactUsLinkComponent
Reverse Transcription PCR (RT-PCR): The Molecular Biology of HIV Replication
Reverse Transcription PCR (RT-PCR): The Molecular Biology of HIV Replication
# de catalogue 470024-524
Fournisseur:  Edvotek
KT RT-PCR:MODEL FORMOLEC BIO OF HIV
Reverse Transcription PCR (RT-PCR): The Molecular Biology of HIV Replication
# de catalogue 470024-524
Fournisseur:  Edvotek
Spécifications
  • Description:
    RT-PCR: Module for Molecular Biology of HIV Replication Kit
  • No. of Lab Groups:
    6
  • Cat. no.:
    470024-524
  • Supplier No.:
    335
Spécifications
A propos de cet article
Modern Application of Advanced Science
  • Advanced technique to explore transcription
  • Time required: ~90 min plus 2 hours for PCR
A specific mRNA is reverse transcribed to double-stranded DNA. This DNA product is then amplified by PCR. This reaction demonstrates the mode of replication of HIV, which contains reverse transcriptase. This experiment is the first introduction of a commercial RNA experiment for the classroom laboratory.
Kit includes: instructions, RNA Template, primer Mix, RT-PCR reaction beads, RNase-free water, DNA size ladder, agarose, electrophoresis buffer, InstaStain® Ethidium Bromide.
Required, but not included: thermal cycler, electrophoresis apparatus, power supply, microcentrifuge, automatic micropipette with tips, balance, microwave or hot plate, UV transilluminator, pipette pump or bulb, lab glassware, distilled or deionized water.
Packaging: Some components require freezer storage on arrival
Customers Who Bought This Also Bought