Spécifications
- Description:RT-PCR: Module for Molecular Biology of HIV Replication Kit
- No. of Lab Groups:6
- Cat. no.:470024-524
- Supplier No.:335
Spécifications
A propos de cet article
Modern Application of Advanced Science
- Advanced technique to explore transcription
- Time required: ~90 min plus 2 hours for PCR
A specific mRNA is reverse transcribed to double-stranded DNA. This DNA product is then amplified by PCR. This reaction demonstrates the mode of replication of HIV, which contains reverse transcriptase. This experiment is the first introduction of a commercial RNA experiment for the classroom laboratory.
Kit includes: instructions, RNA Template, primer Mix, RT-PCR reaction beads, RNase-free water, DNA size ladder, agarose, electrophoresis buffer, InstaStain® Ethidium Bromide.
Required, but not included: thermal cycler, electrophoresis apparatus, power supply, microcentrifuge, automatic micropipette with tips, balance, microwave or hot plate, UV transilluminator, pipette pump or bulb, lab glassware, distilled or deionized water.