Round Bug Viewer
# de catalogue 470004-762
Fournisseur: EDUCATIONAL INSIGHTS, INC.
Spécifications
- Magnification:3×
- Description:Viewer, Bug, Round, 73 mm, 3"
- Cat. no.:470004-762
- Supplier No.:EI-5111
Spécifications
A propos de cet article
Provide students with a well-rounded view of insect life with these well-rounded viewers.
Viewers are 73 mm in diameter and feature a high quality magnifier lid and air holes.
Customers Who Bought This Also Bought