ContactUsLinkComponent
Rotary Engine Model
Rotary Engine Model
# de catalogue 470121-114
Fournisseur:  VWR International
MODEL ROTARY ENGINE
Rotary Engine Model
# de catalogue 470121-114
Fournisseur:  VWR International
Spécifications
  • Description:
    Rotary Engine Model
  • Cat. no.:
    470121-114
  • Supplier No.:
    PH0496
Spécifications
A propos de cet article
Close-Up Cross Section.
  • Provide visual representation of the principle operation of rotary engines
  • Model is mounted on base
The power piston in this model is an arch-like triangular rotar, which generates an epitrechoid on rotation. So you can view internal construction details, the model is cut away for easy observation.
Customers Who Bought This Also Bought