Ward's® Rock Thin Section
# de catalogue 470106-374
Fournisseur: Avantor
Spécifications
- Description:Rock Thin Section Set/6
- Cat. no.:470106-374
- Supplier No.:WARD470106-374
Spécifications
A propos de cet article
Use these thin-section slides with a polarizing microscope to view rock specimens in detail. Includes: biotite granite, olivine basalt, red sandstone, encrinal limestone, mica schist, augen gneiss, storage box and identification guide.
Note: collections are subject to change depending upon availability of individual items. Set of six.
Customers Who Bought This Also Bought