ContactUsLinkComponent
Ward's® Restriction Mapping of DNA Lab Activity
Ward's® Restriction Mapping of DNA Lab Activity
# de catalogue 470030-388
Fournisseur:  Avantor
RESTRICTION MAPPING OF DNA GEL KIT
Ward's® Restriction Mapping of DNA Lab Activity
# de catalogue 470030-388
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Restriction Mapping of DNA Kit
  • No. of Lab Groups:
    8
  • Cat. no.:
    470030-388
Spécifications
A propos de cet article
Perform an Important Technique Used to Locate Genes on Chromosomes
  • Practice restriction mapping
  • Materials for 8 lab groups
  • Time Required: ~120 minutes
To locate a particular gene, scientists first identify landmarks along a chromosome or a long stretch of DNA. The most important landmarks of a physical map are the cleavage, or restriction, sites of restriction enzymes. A chromosome “map” specifies the landmarks and the distances between them, measured in nucleotides; the ultimate physical map is an exact order of the nucleotides created with a process known as DNA sequencing. In this lab activity that can be completed in two 60 minutes sessions, students electrophorese lambda DNA digested with restriction enzymes EcoR I and Hind III, analyze the fragment patterns and their sizes, and construct a chromosome map. The kit includes enough materials to run eight gels, a teacher’s guide, and student copymaster.
Materials required, not included: An electrophoresis chamber, power supply, and 1-20 μL micropipettes.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Customers Who Bought This Also Bought