Resonance Tubes
Fournisseur: ROCHESTER APPAREL & AD SPECIALTIES
Certificats
A propos de cet article
Perfect for any number of resonance experiments
- Available in two sizes:
- 11 7/8" x 1"
- 17 3/4" x 1 1/2"
Durable resonance tubes are made of glass and feature annealed ends.
Customers Who Bought This Also Bought