Rescue Critters® K-9 Thoracentesis Manikin
# de catalogue 470126-196
Fournisseur: THALES & CO.
Spécifications
- Description:K-9 Thoracentesis
- Cat. no.:470126-196
- Supplier No.:925
Spécifications
A propos de cet article
Emergency Response Training For Vets
- Convenient anatomical landmarks
- Perform chest tube placements
- Made to order
This carefully crafted canine Thoracentesis manikin will help practitioners in performing chest tube placements, and simulating emergency trauma by aspirating air and fluid from the thoracic cavity. Convenient anatomical landmarks are placed at key points for ease of use and quick learning.
Ordering information: This item is made to order. Returns cannot be accepted.
Customers Who Bought This Also Bought