ContactUsLinkComponent
Red Cabbage pH Indicator Kit
Red Cabbage pH Indicator Kit
# de catalogue 470145-176
Fournisseur:  Universe of Science
RED CABBAGE PH INDICATOR KIT
Red Cabbage pH Indicator Kit
# de catalogue 470145-176
Fournisseur:  Universe of Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Red Cabbage pH Indicator Kit
  • Cat. no.:
    470145-176
  • Supplier No.:
    2150-B
Spécifications
A propos de cet article
Use Red Cabbage Extract as a Natural Acid-Base Indicator
No need to be intimidated by acid-base reactions with this classroom safe pH activity. Use red cabbage extract, a natural acid-base indicator, to illustrate common reactions. Show students how to use pH charts and distinguish between chemical components. Made of all food grade materials, clean up and disposal of this project is quick and easy. Materials in this kit include 20 g red cabbage extract, plastic beaker, pipet, pH chart, and instructions.
Customers Who Bought This Also Bought