Ward's® Preserved Rats
Supplier: Ward's Science
Certificates
About this item
Formalin preserved rats. Available in vacuum packs or pails.
- Mammalian anatomy
- General dissection
- High quality preservation
Our rat specimens are carefully preserved in an extended position and are mature subjects for students to examine fully developed animals, their organ systems, and their skeletal, muscular, and reproductive structures. We carry plain preserved, single, double, and triple injected rats. Injected rats are made without incisions in the neck, ensuring the specimens are intact. Sizes 7 to 9”.
Customers Who Bought This Also Bought