Rare Earth Magnet
# de catalogue 470326-954
Fournisseur: PICOSOLUTIONS GROUP INC SE
Spécifications
- Description:Rare earth magnet, pack of 4
- Cat. no.:470326-954
Spécifications
A propos de cet article
Compatible with Variety of Windmill Kits
- Strength 0.395 Tesla/3,950 Gauss
- Size 1.875×0.875×0.375" thick
- Pole marked with adhesive sticker
This ceramic, grade 5 magnet is ideal for use in constructing the alternator used in a variety of wind turbine kits. Use caution: ceramic magnets are magnetically strong, but physically fragile and can chip or crack easily.
Caution: WARNING! These magnets are strong and can pinch your fingers if not handled carefully. Persons wearing pacemakers should not handle strong magnets.
Customers Who Bought This Also Bought