Ward's® Rainfall Simulator
# de catalogue 470029-378
Fournisseur: Plas-Labs
Spécifications
- Description:WARD’S Rainfall Simulator
- Cat. no.:470029-378
- Supplier No.:152-RM
Spécifications
A propos de cet article
For More Realistic Precipitation
With the ready-to-use model, molded of heavy-duty plastic with support rods, you can produce a gentle rain or a down-pour. Designed for use with WARD’S stream tables. Size: 17"L x 8 1/2"W x 2"H.
Customers Who Bought This Also Bought