Radiometer
# de catalogue 470148-644
Fournisseur: VWR International
Spécifications
- Description:Radiometer
- Cat. no.:470148-644
- Supplier No.:WL1734
Spécifications
A propos de cet article
This simple apparatus demonstrates radiant heat.
- Diameter: 3"
- Height: 5"
- Vanes Turn When Dark Sides Absorb Heat
Vanes on the rotating shaft are light on one side but dark on the other.
When the dark sides of the vanes absorb light, the vanes turn.
Customers Who Bought This Also Bought