ContactUsLinkComponent
Ward's® QUIKView DNA Stain for Gel Electrophoresis
Ward's® QUIKView DNA Stain for Gel Electrophoresis
# de catalogue 470006-426
Fournisseur:  Edvotek
STAIN ELECTROPHOR QUICKVIEW DNA 60 ML
Ward's® QUIKView DNA Stain for Gel Electrophoresis
# de catalogue 470006-426
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    WARD’S QUIKView DNA Stain for Gel Electrophoresis
  • Cat. no.:
    470006-426
  • Supplier No.:
    609-WARDS
Spécifications
A propos de cet article
Stains and Destains in 45 Minutes, Providing Unsurpassed Visibility
Fast acting, completely safe, highly sensitive, and versatile, our DNA stain is a great alternative to the common methylene blue stain. WARD’S exclusive DNA stain works quickly, requiring only 20–30 minutes to stain a gel, and approximately 15 minutes for destaining. QUIKView destains in water, providing a totally clear background that highlights DNA banding patterns, making them visible even without a light source such as a light table. You can also incorporate the stain into the gel matrix and buffer to stain the gel during electrophoresis. QUIKView detects as low as 0.3 μg of DNA. And, once the gels are stained, they can be stored for months without fading.
Quickview stain -- product reformulation-- now a 10x concentration, after diluting the final volume is 600mL which is enough for 30 gels.
Customers Who Bought This Also Bought