ContactUsLinkComponent
Qualitative ELISA Kit
Qualitative ELISA Kit
# de catalogue 470120-906
Fournisseur:  Edvotek
QUALITATIVE ELISA KIT
Qualitative ELISA Kit
# de catalogue 470120-906
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Qualitative ELISA Kit
  • Cat. no.:
    470120-906
  • Supplier No.:
    278-B
Spécifications
A propos de cet article
Demonstrate the quantitation of varying concentrations of viral antigens as detected by the intensity of the color reaction due to the accumulation of products.
  • Material for Six Groups with 12 Reactions Each
  • Instructions with Background Information Included
  • Some Components Require 4°C Refrigeration
The kit provides material for six groups with 12 reactions each.
Some components require 4°C refrigeration upon receipt.
Ordering information: This kit includes antigens, antibodies, and secondary antibodies; substrate solution; phosphate buffered saline; blocking agent; stop solution; microtiter plates; transfer pipets; micro-test tubes; and instructions with background information. The kit requires, but does not include, distilled or deionized water, incubation oven, and automatic pipets (5—50 µL) and tips.
Delivery: This item is shipped ambient.
Customers Who Bought This Also Bought