Pyrite Mineral Display, Large
# de catalogue 470154-900
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Pyrite Mineral Display, 1" x 2".
- Cat. no.:470154-900
- Supplier No.:3494231
Spécifications
A propos de cet article
Spain
Distinctive and well-formed cubic crystal intergrowths of bright golden pyrite. Average size: 1" x 2". Limited Edition. Spain.
Customers Who Bought This Also Bought