ContactUsLinkComponent
Purification and Size Determination of GFP/BFP Kit (EDVO-Kit 255)
Purification and Size Determination of GFP/BFP Kit (EDVO-Kit 255)
# de catalogue 470112-846
Fournisseur:  Edvotek
PURIF AND SIZE DETER OF GFP
Purification and Size Determination of GFP/BFP Kit (EDVO-Kit 255)
# de catalogue 470112-846
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Purification and Size Determination of GFP/BFP Kit
  • Cat. no.:
    470112-846
  • Supplier No.:
    255-B
Spécifications
A propos de cet article
Students use Green Fluorescent Protein (GFP) to identify the localization and function of proteins in cells.
  • Material for Six Groups Sharing Three Gels
  • Fluorescent Property of GFP Functions as Assay
  • GFP Tags Proteins in Cells
  • Optional Activity Included
  • Some Components Require -20°C Storage
GFP has been isolated and cloned from the jellyfish Aequorea victoria for this kit. The protein gives a green fluorescent glow under a long UV light, and it has been used as a tag to follow proteins in cells. In this experiment, the unique fluorescent property of GFP is used as an assay during its purification from an E. coli extract. Column fractions that contain GFP are identified by green fluorescence. As an optional activity, purified protein fractions can be submitted to denaturing SDS polyacrylamide gel analysis to estimate the purity and size of the GFP protein.
This kit includes material for six groups sharing three gels. Vertical gel electrophoresis apparatus, a power supply, 5—50 μL micropipets, and polyacrylamide gels are required but not included.
Delivery: The kit is shipped at room temperature, but some components require -20°C storage upon receipt.
Customers Who Bought This Also Bought