ContactUsLinkComponent
Pulse Glass
Pulse Glass
# de catalogue 470146-174
Fournisseur:  American Scientific
Small Business Enterprise
PULSE GLASS
Pulse Glass
# de catalogue 470146-174
Fournisseur:  American Scientific
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Pulse Glass
  • Cat. no.:
    470146-174
  • Supplier No.:
    2010-20
Spécifications
A propos de cet article
Ideal for demonstrating that vapor pressure increases when temperature increases.
Ideal for demonstrating that vapor pressure increases when temperature increases.
Two glass bulbs are connected with a glass tube, and contain a colored, volatile liquid. Holding one of the bulbs in your hand causes the evaporation of a portion of the liquid that forces the liquid to the other bulb. Length is 23.5 cm.
  • Easy to use
  • Length 23.5 cm
Two glass bulbs are connected with a glass tube, and contain a colored, volatile liquid. Holding one of the bulbs in your hand causes the evaporation of a portion of the liquid that forces the liquid to the other bulb.
Customers Who Bought This Also Bought