Protist Motility Slide
Catalog # 470177-488
Supplier: Avantor
Specifications
- Description:Protist Motility
- Cat. no.:470177-488
- Stain:Fast Green and Mallory's, Trichrome
- Supplier No.:WARD470177-488
- Orientation:Explano-Mount™
Specifications
About this item
Illustrates How Organisms Move in their Environment
- Identifying features clearly distinguishable
- Rigorous quality control standards
Three classes represented including: Amoeba, Euglena, and Paramecium.
Customers Who Bought This Also Bought