A propos de cet article
Isolated from sewage, human intestines, and urinary tract infections. Spreading colonies are seen on agar growth medium.
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Motility: Positive
- Morphology: Rods
- Gram Reaction: Negative
- Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout