Protein InstaStain®
# de catalogue 470002-532
Fournisseur: Edvotek
Numéro CAS:
Spécifications
- Description:Protein InstaStain®, For 15 Gels
- Cat. no.:470002-532
- Supplier No.:J
Spécifications
A propos de cet article
High Quality Results, Excellent for Photodocumentation
Stain protein gels in just 20 minutes, plus fixing and destaining times. Protein InstaStain® is far faster than liquid blue stains and ensures a yield of uniform gel results. Environmentally friendly InstaStain® also uses a solid matrix to avoid large amounts of liquid waste. For 15 gels.
Customers Who Bought This Also Bought