ContactUsLinkComponent
Projectile Launcher
Projectile Launcher
# de catalogue 470014-320
Fournisseur:  VWR International
DEMONSTRATION PROJECTILE LAUNCHER
Projectile Launcher
# de catalogue 470014-320
Fournisseur:  VWR International
Spécifications
  • Description:
    Projectile Launcher
  • Supplier No.:
    PH0343
Spécifications
A propos de cet article
This launcher is easy to assemble, and it delivers consistent classroom results.
  • Rugged and Durable Construction
  • Easy Assembly
  • Consistent Classroom Results
  • Spring-Loaded Gun Design
  • Instructions Included
It's built to withstand the rigors of any hands-on lab and precision machined to give the same result every time.
The range is variable up to 10', and it measures any angle up to 90° within 1/2° accuracy with the plumb bob.
Ordering information: This kit includes an assembled spring-loaded gun, two 1" dia. balls (one steel, one aluminum), 4 experiments, protractor attachment, plumb bob, brace, and instructions.
Customers Who Bought This Also Bought