Prisms
Catalog # 470149-510
Supplier: VWR International
Specifications
- Description:Prisms, Acrylic Equilateral Prism, 75mm sides, 9mm thick
- Cat. no.:470149-510
- Supplier No.:P30273
Specifications
About this item
Clear, Short Prisms for Index of Refraction and Dispersion
Equilateral prisms are clear with long sides for refractive index measurements, trapezoidal prisms have a frosted base to show ray paths inside the prism.
Customers Who Bought This Also Bought