ContactUsLinkComponent
Preserved Specimen Conditioning Fluid
Preserved Specimen Conditioning Fluid
# de catalogue 470024-842
Fournisseur:  Avantor
Numéro CAS:  
WARDS PM SPECIMEN CONDITIONR. SOL 500 ML
Preserved Specimen Conditioning Fluid
# de catalogue 470024-842
Fournisseur:  Avantor
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Conditioning Fluid, Laboratory, 500 ml Spray Bottle
  • Cat. no.:
    470024-842
  • Grade:
    Laboratory
  • Supplier No.:
    WARD470024-842
Spécifications
A propos de cet article
Hazard Info: Toxic
Shelf Life (months): 36
Storage: Blue
This solution can be used as a mold inhibitor for dissection specimens if you choose to dissect your specimen over multiple days/weeks.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Customers Who Bought This Also Bought