Prepared Electrophoresis Agar
Fournisseur: Ward's Science
Certificats
A propos de cet article
Jump to the electrophoresis step quicker - let us prepare your gel for you.
- Convenient, just melt and pour.
- Choose the concentration to view DNA fragments from 0.1 to 35 kb in length
2.0% Agarose is used to resolve DNA fragments in the range of 0.1 to 5kb in length.
0.8% Agarose is used to resolve DNA fragments in the range of 0.6-35kb in length.
Customers Who Bought This Also Bought