Potato Dextrose Agar
# de catalogue 470193-232
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Description:Dehydrated Potato Dextrose Agar
- Cat. no.:470193-232
- Supplier No.:880007
Spécifications
A propos de cet article
For cultivation of fungi and yeasts.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 .
Customers Who Bought This Also Bought