Portable Fuming Chamber
# de catalogue 470019-340
Fournisseur: FORENSICS SOURCE
Spécifications
- Description:Portable Fuming Chamber
- Size:4 lbs.
- Cat. no.:470019-340
- Dimensions:15W×8D×11" H
- Supplier No.:1-4676
Spécifications
A propos de cet article
Bring Your Equipment to the Crime!
- Made from durable ABS plastic
- Includes (2) hanging bars with (4) clips for suspending evidence
- Magnetic latch simplifies chamber loading and unloading
- Convenient metal carrying handle allows for easy transportation and storage at the crime scene or the lab
The roomy chamber fits a wide variety of commonly processed evidence including cans, bottles and weapons. A special port incorporated in the design allows the attachment of an exhaust tube to vent fumes. This small but capable chamber can be used at home, in the lab, or in the field!
Customers Who Bought This Also Bought