Porphyridium
# de catalogue 470179-906
Fournisseur: Ward's Science
Spécifications
- Description:Porphyridium (Small)
- No. of Students:30
- Cat. no.:470179-906
- Packaging:Tube
- Media:Erdschreiber's medium
- Supplier No.:WARD470179-906
Spécifications
A propos de cet article
Marine. Unicellular. Erdschreiber’s medium.
Characterized by reddish plastids (rhodoplasts).
Customers Who Bought This Also Bought