Porcelain Evaporating Dishes
Catalog # 470020-006
Supplier: GSC International
Specifications
- Capacity:75 ml
- Height:32 mm
- Cat. no.:470020-006
- Outer Diameter:82 mm
- Supplier No.:7-525-2
Specifications
About this item
Evaporating basin designed to evaporate excess solvents to produce a concentrated solution or a precipitate.
- Each dish features pour spout
- Glazed inside and out, except for bottom surface
- Easily wipe clean the inside
Designed for use in the laboratory, but can be used in the home as well or even in a set for 'potions'.
Customers Who Bought This Also Bought