Laboratory Tank Starter Innocula
Fournisseur: Avantor
Certificats
A propos de cet article
Samples from our well-established aquculture systems will help get your systems up and running.
- Nonsterile
- Contains beneficial bacteria and algae
- Jump-start you tank acclimation process.
Pulled from either our freshwater or saltwater systems these water samples contain nutrients and various beneficial bacteria and algae that will establish themselves in your classroom tanks and begin building up your tank's important natural flora.
Customers Who Bought This Also Bought