Polysiphonia
Fournisseur: Avantor
Certificats
A propos de cet article
Marine. Filamentous; polysiphonious construction. Collected.
Characterized by reddish plastids (rhodoplasts).
Customers Who Bought This Also Bought
Marine. Filamentous; polysiphonious construction. Collected.
Characterized by reddish plastids (rhodoplasts).