Polypropylene Stirring Rod
Catalog # 30620-402
Supplier: Dynalon
Specifications
- Description:Polypropylene Stirring Rod
- Cat. no.:30620-402
- Supplier No.:212455
Specifications
About this item
The plastic rod stirs silently and the flattened end may also be used as small scoop or spatula.
¹/₄ inch diameter and 9¹/₂ inch length.
Customers Who Bought This Also Bought