Polypropylene Short Stem Funnels, 60°, Bel-Art
# de catalogue 470145-358
Fournisseur: Bel-Art
Spécifications
- Capacity:91.8 ml
- Cat. no.:470145-358
- Top I.D.:75 mm
- Stem Length:75 mm
- Fits Paper Diameter:125 mm
- Supplier No.:F14676-0075
Spécifications
A propos de cet article
The external ribbing on this funnel prevents air lock.
- Funnel Has 60° Angle
- Internal Ribs for Speedier Filtrations
- External Ribbing Prevents Air Lock
- Autoclavable
The funnel has a 60° angle. The internal ribs allow for speedier filtrations.
It is autoclavable.
Customers Who Bought This Also Bought