ContactUsLinkComponent
Graduated Polypropylene Pitchers
Graduated Polypropylene Pitchers
# de catalogue 470201-328
Fournisseur:  VWR International
Graduated Polypropylene Pitchers
Graduated Polypropylene Pitchers
# de catalogue 470201-328
Fournisseur:  VWR International
Log in for availability
Select an Option
3000 ml
Select an Option
100 ml
Select an Option
470201-328
Select an Option
A10353/6
A propos de cet article
These pitchers are made with shatter-proof polypropylene.
  • Autoclavable up to 135°C
  • Resistant to Most Acids, Bases, Alcohol, Ethers, Oils, and Hydrocarbons
  • Easy-to-See Graduations
  • Shatter-Proof Polypropylene Construction
Autoclavable up to 135°C, the pitchers are resistant to alcohol, ethers, hydrocarbons, oils, bases, and most acids.
They're marked with well-defined graduations.
Customers Who Bought This Also Bought