Large Utility Funnels, Hutzler
# de catalogue 470177-822
Fournisseur: Hutzler Manufacturing
Spécifications
- Length:60 mm
- Description:Large Utility Funnels, Pack of 6
- Material:Polyethylene
- Cat. no.:470177-822
- Inner Diameter:160 mm
- Supplier No.:832-WN
Spécifications
A propos de cet article
These large utility funnels will withstand temperatures up to 250°F (121°C).
Customers Who Bought This Also Bought