Polypropylene Beakers with Handles
Catalog # 470144-838
Supplier: United Scientific Supplies
Specifications
- Capacity:1000 ml (34 oz.)
- Description:Polypropylene Beakers with Handles, Tall Form, Capacity: 1000 mL
- Cat. no.:470144-838
- Supplier No.:81103
Specifications
About this item
These inexpensive pitcher-style beakers are chemical resistant and have permanent graduations in liters and milliliters. Made of autoclavable polypropylene.
- Easy grip handle
- Raised graduations
- Autoclavable
Large scale beaker with handle to make pouring easy.
Customers Who Bought This Also Bought